Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-Cytosolic Sulfotransferase 1A3/SULT1A3/CSPS, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP131809

Catalog No. NBP131809


Add to cart

Description

Description

Cytosolic Sulfotransferase 1A3/SULT1A3/CSPS Polyclonal antibody specifically detects Cytosolic Sulfotransferase 1A3/SULT1A3/CSPS in Human samples. It is validated for Western Blot.
Specifications

Specifications

Cytosolic Sulfotransferase 1A3/SULT1A3/CSPS
5.96mg/mL
Western Blot 1:500-1:3000
Purified
SULT1A3
Recombinant protein encompassing a sequence within the center region of human SULT1A3. The exact sequence is proprietary.
34 kDa
0.1 ml
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Polyclonal
Tested in human. Predicted cross-reactivity based on sequence identity: Monkey (94%). Not yet tested in any other species.
Western Blot
Unconjugated
PBS (pH 7.0), 1.0% BSA and 20% Glycerol with 0.01% Thimerosal
EC 2.8.2, EC 2.8.2.1, HAST, M form) phenol sulfotransferase, MGC117469, Monoamine-sulfating phenol sulfotransferase, ST1A3/ST1A4, sulfotransferase family, cytosolic, 1A, phenol-preferring, member 3, Sulfotransferase, monoamine-preferring, Thermolabile phenol sulfotransferase
Rabbit
IgG
Immunogen affinity purified
RUO
Primary
6818
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only