Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
D-dimer Mouse anti-Human, Clone: DD5, Invitrogen™
Mouse Monoclonal Antibody
Supplier: Thermo Scientific MA120030
Description
MA1-20030 detects D-Dimer in human samples. MA1-20030 has been successfully used in ELISA and Western blot procedures. The MA1-20030 immunogen is homogenized fibrin clot.
Fibrinogen is the main protein of blood coagulation system. It consists of two identical subunits that contain three polypeptide chains: alpha, beta and gamma. The process of blood coagulation results in the activation of fibrinogen into fibrin by thrombin and fibrin polymerization. Fibrin clot is then digested by plasmin, and fibrin degradation products of different molecular weights are released into the bloodstream.Specifications
D-dimer | |
Monoclonal | |
Unconjugated | |
P02671 | |
Mouse | |
Protein A | |
RUO | |
2243 | |
4° C | |
Liquid |
ELISA, Western Blot | |
DD5 | |
FGA | |
FGA | |
Homogenized fibrin clot. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2b |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title