Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-D31, Clone: RM0032-1D12, Abcam™ Non-distribution product as customer accommodation.

Manufacturer:  Abcam AB56299100UG

Catalog No. NC0054389


Add to cart

Description

Description

Host Species: Rat
Clone: RM0032-1D12
Isotype: IgG2a
Cross Reactivity: Mouse
Immunogen: Tissue / cell preparation: Mouse endothelial membrane protein

Applications:

Immunohistochemistry (Paraffin)

Specifications

Specifications

D31
0.5mg/mL
Purified
Tissue / cell preparation: Mouse endothelial membrane protein
100μg
Primary
Murine
RM0032-1D12
Unconjugated
Rat
IgG2a
Cancer and Tumor Biology, Signal Transduction, Stem Cell Biology
Monoclonal
Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit