Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-D4-GDI/RhoGDI2, Polyclonal, Novus Biologicals™

Manufacturer:  Novus Biologicals NBP132463

Catalog No. NBP132463

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Specifications

D4-GDI/RhoGDI2
1.0mg/mL
Western Blot 1:500-1:3000
Affinity Purified
ARHGDIB
Recombinant protein encompassing a sequence within the center region of human Rho GDI2. The exact sequence is proprietary.
23 kDa
100 ul
Apoptosis, Signal Transduction
Primary
397
Human, Mouse
Western Blot
Unconjugated
0.1M Tris (pH 7.0), 0.1M Glycine and 10% Glycerol with 0.01% Thimerosal
ARHGDIB Rho GDP dissociation inhibitor (GDI) beta D4, GDIA2, GDID4, Ly-GDI, LYGDI, RAP1GN1, RhoGDI2
Rabbit
IgG
Immunogen affinity purified
RUO
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Polyclonal
Tested in human. Expected to cross react based on sequence identity: Mouse (90%), Rat (89%), Xenopus laevis (81%), Bovine (93%).
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only