Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DAAM1 Rabbit anti-Bovine, Canine, Equine, Human, Mouse, Rat, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA523222

Catalog No. PIPA523222


Add to cart

Description

Description

DAAM1 is a diaphanous-related formin first studied as a novel Dishevelled binding protein and shown to be crucial for the planar cell polarity pathway in Xenopus. DAAM1, like other formins, directs nucleation and elongation of new actin filaments using its conserved formin-homology-2 (FH2) domain. It is a Rho-regulated formin implicated in neuronal development and regulation of gastrulation. It was identified as a binding protein of the Wnt receptor-associated protein Dishevelled. It also binds the GTP-bound RhoA and increases its GTP-bound form population in cells. It exists in an autoinhibited state via intramolecular interactions between its amino-terminal GBD and carboxyl-terminal DAD domains. Structurally the protein includes a GTPase-binding domain (GBD), a diaphanous inhibitory domain (DID), and a coiled-coil region followed by the FH1, FH2 and C-terminal diaphanous auto-regulatory domain (DAD). In humans, the DAAM1 gene is mapped to chromosome 14q23.1.
Specifications

Specifications

DAAM1
Polyclonal
Unconjugated
DAAM1
Liquid
DAAM1
Synthetic peptide (18 FRNNDHPEITYRLRNDSN 35) of DAAM1 protein.
Protein A
RUO
Antibody
Polyclonal
Bovine, Canine, Equine, Human, Mouse, Rat
Western Blot
0.5 mg/mL
PBS with 0.05% BSA and 0.05% sodium azide
Q9Y4D1, Q8BPM0
disheveled-associated activator of morphogenesis 1
Rabbit
IgG
100 μg
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
100051203, 208846, 23002, 314212, 490714, 513742
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit