Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DAB1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 1 publication
Supplier: Novus Biologicals NBP216095
Description
DAB1 Polyclonal specifically detects DAB1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
DAB1 | |
Polyclonal | |
Unconjugated | |
PBS, 20% Glycerol (pH7) with 0.025% Proclin 300 | |
disabled (Drosophila) homolog 1, disabled homolog 1, disabled homolog 1 (Drosophila) | |
Rabbit | |
64 kDa | |
0.1 mL | |
Growth and Development, Neuroscience, Signal Transduction | |
1600 | |
Human, Mouse, Rat | |
IgG |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
1mg/mL | |
Western Blot 1:500-1:3000, Immunohistochemistry 1:100-1:1000, Immunocytochemistry/Immunofluorescence 1:100-1:1000, Immunohistochemistry-Paraffin 1:100-1:1000 | |
O75553 | |
DAB1 | |
Recombinant protein encompassing a sequence within the N-terminus region of human DAB1. The exact sequence is proprietary. | |
Affinity Purified | |
RUO | |
Primary | |
Human, Mouse. Expected to cross react based on sequence identity: Pig (100%), Rat (100%). | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only