Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DAB1 Sheep anti-Mouse, Polyclonal, Invitrogen™

Sheep Polyclonal Antibody

Manufacturer:  Invitrogen PA112425

Catalog No. 01-674-142


Add to cart

Description

Description

A suggested positive control for this product is 293 cells transfected with Dab-1 HA tag.

Disruption of Dab1 results in aberrant migration of neurons during development and disorganization of laminar structures throughout the brain. Dab1 is thought to function as an adaptor molecule in signal transduction processes. It contains a protein-protein interaction (PI) similar to the phosphotyrosine binding domain of the Shc oncoprotein, it is phosphorylated by the Src protein tyrosine kinase, and it binds to SH2 binding domains in a phosphotyrosine dependent manner. DAB1 acts downstream of reelin in a signalling pathway.
Specifications

Specifications

Dab1
Polyclonal
PBS with 0.08% sodium azide
P97318
RP23-356E21.1, AI956902, C630028C02Rik, scm, scr, scrambler, yot
Sheep
IgG
250 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
13131
Western Blot
Unconjugated
Dab1
Liquid
DAB1
residues 1-179 of mouse disabled coupled to a GST fusion protein.
Ammonium sulfate precipitation
RUO
Antibody
Polyclonal
Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit