Learn More
anti-Dact1, Polyclonal, Novus Biologicals™
Manufacturer:  Novus Biologicals NBP141014
Catalog No. NBP141014
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications
Specifications
Dact1
Unconjugated
PBS (pH 7.4) with 0.09% Sodium Azide
Purified
DACT1
A peptide corresponding to amino acids 373-386 within exon4 of mouse Dact1.
IgG purified
RUO
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Polyclonal
This antibody recognises mouse Dact1 (Dapper homolog 1), an intracellular protein and member of the dapper family, which acts as a positive regulator of the signalling pathways of Dishevelled 2 (DVL2), a cytoplasmic protein which plays a pivotal role in Wnt signalling during vertebrate development.
Western Blot
Western Blot 1:100-1:2000
Q8R4A3
DAPPER, dapper, antagonist of beta-catenin, homolog 1 (Xenopus laevis), DAPPER1dapper homolog 1, antagonist of beta-catenin (xenopus), DPR1dapper homolog 1, antagonist of beta-catenin, FRODOdapper homolog 1, HDPR1, Hepatocellular carcinoma novel gene 3 protein, heptacellular carcinoma novel gene 3, HNG3, THYEX3
Rabbit
IgG
0.05 mg
Cancer
Primary
51339
Mouse