Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DACT1 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA525814

Catalog No. PIPA525814

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Specifications

DACT1
Polyclonal
PBS with 0.09% sodium azide
Q9NYF0, Q8R4A3
DAPPER, DAPPER1, DPR1, FRODO, HDPR1, THYEX3, dapper homolog 1, hepatocellular carcinoma novel gene 3 protein, heptacellular carcinoma novel gene 3
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
51339, 59036
Flow Cytometry, Immunohistochemistry, Western Blot
Unconjugated
DACT1
Liquid
DACT1
KLH conjugated synthetic peptide between 13-41 amino acids from the N-terminal region of human DACT1
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit