Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-DAF-3, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

$119.60 - $314.00

Specifications

Host Species Rabbit
Isotype IgG
Immunogen A synthetic peptide made to the C-terminus (within residues 700-796) of the C.elegans DAF-3 protein. [UniProt# O17532]
Antigen DAF-3
Monoclonal or Polyclonal Polyclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB1001924SS
View Documents
Novus Biologicals
NB1001924SS
0.025 ml
Each for $119.60
Add to cart
 
NB1001924
View Documents
Novus Biologicals
NB1001924
0.1 ml
Each for $314.00
Add to cart
 
Description

Description

DAF-3 Polyclonal antibody specifically detects DAF-3 in C. elegans samples. It is validated for Western Blot, Chromatin Immunoprecipitation, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin.
Specifications

Specifications

Rabbit
A synthetic peptide made to the C-terminus (within residues 700-796) of the C.elegans DAF-3 protein. [UniProt# O17532]
Polyclonal
Primary
Western Blot, ChIP assay, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin)
O17532
Whole antisera with 0.05% Sodium Azide
IgG
DAF-3
Antisera
RUO
C. elegans.
180431
SDS
Product Certifications

For Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit