Learn More
DAP Mouse anti-Human, Clone: 3C5, Invitrogen™
Mouse Monoclonal Antibody
Manufacturer:  Invitrogen MA518699
Catalog No. PIMA518699
Add to cart
Description
Description
Peptide Sequence: MSSPPEGKLE TKAGHPPAVK AGGMRIVQKH PHTGDTKEEK DKDDQEWESP SPPKPTVFIS GVIARGDKDF PPAAAQVAHQ KPHASMDKHP SPRTQHIQQP RK
This gene encodes a basic, proline-rich, 15-kD protein. The protein acts as a positive mediator of programmed cell death that is induced by interferon-gamma.
Specifications
Specifications
DAP
Monoclonal
0.2-1.0 mg/mL
PBS with no preservative; pH 7.4
P51397
DAP, Death-associated protein 1
Mouse
IgG2a, kappa
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
1611
ELISA, Western Blot
3C5
Unconjugated
DAP
Liquid
DAP
DAP (AAH02726, 1 a.a. ∽ 102 a.a) full-length recombit protein with GST tag. MW of the GST tag alone is 26 KDa.
Affinity chromatography
RUO
Antibody
Monoclonal
Human