Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DAP Kinase Mouse anti-Human, Unlabeled, Clone: 17, BD

Mouse Monoclonal Antibody

Manufacturer:  BD Biosciences 610290

 View more versions of this product

Catalog No. BDB610290


Add to cart

Description

Description

Chronic exposure to extracellular signals such as interferons induce the inhibition of cell proliferation followed by cell death. The gene for DAP Kinase (Death Associated Protein Kinase) was identified using a novel approach named Technical Knockout. Briefly, Hela cells were transfected with an antisense cDNA expression library, then exposed to Interferon-γ. The surviving cells, with their antisense cDNAs, were rescued and the “protecting” genes isolated. The DAP Kinase gene encodes a protein of 1423 amino acids, a molecular weight of 160kDa, a kinase domain at its amino terminal region, ankyrin repeats in the middle region, and a death domain at the extreme C-terminus. DAP Kinase phosphorylates at Ser/Thr residues in a Ca2+/Calmodulin-dependent fashion. It has been demonstrated that Ca2+/Calmodulin binds directly to DAP Kinase at its amino terminal region. In addition, immunostaining studies localized DAP Kinase in association with the actin filaments where it may phosphorylate myosin light chain. Thus, this novel cytoskeletal and Ca2+/Calmodulin-dependent protein kinase plays a role in interferon-γ-induced cell death.

Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blotting

Specifications

Specifications

DAP Kinase
17
Unconjugated
Affinity Purified
Aqueous buffered solution containing BSA
50μg
Cell Biology
Primary
Human
Western Blot
250μg/mL
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Murine
IgG1
RUO
Store undiluted at -20°C.
Monoclonal
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only.