Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DAP5 Mouse anti-Human, Mouse, Clone: 39C534.1, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA141094

Catalog No. 01-676-465


Add to cart

Description

Description

Suggested positive control: Jurkat, antigen standard for EIF4G2 (transient overexpression lysate), Jurkat whole cell lysate.

Apoptosis, or programmed cell death, occurs during normal cellular differentiation and development of multicellular organisms. Recently, five novel apoptotic genes (named DAP genes for: Death Associated Proteins) have been identified that code for proteins, which display a diverse spectrum of biochemical activities. These include a novel type of calcium/calmodulin-regulated kinase, which carries ankyrin repeats and a death domain (DAP-kinase), a nucleotide-binding protein (DAP-3), a small proline-rich cytoplasmic protein (DAP-1), and a novel homolog of the eIF4G translation initiation factor (DAP-5) (1-2). DAP-5 (also named p97 and NAT1) is a ubiquitously expressed member of the translation initiation factor eIF4G family that lacks the eIF4E binding site. DAP-5 was cloned from HeLa cells using a functional approach in attempt to isolate cDNA fragments, which convey resistance to gamma interferon (IFN-g)-induced programmed cell death (3). A fragment of the cDNA when expressed as a mini-protein of 28 kDa protects the cell from IFN-g induced apoptosis. The full-length DAP-5 cDNA codes for a protein of approximate molecular weight of 97 kDa. Low level of expression protects the cells, whereas high level of expression results in cell death (3-4).
Specifications

Specifications

DAP5
Monoclonal
0.5 mg/mL
PBS with 0.2% gelatin and 0.05% sodium azide
P78344, Q62448
AAG1, DAP5, FLJ41344, NAT1, P97
Mouse
IgG1
100 μg
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
13690, 1982
Flow Cytometry, Western Blot
39C534.1
Unconjugated
EIF4G2
Liquid
EIF4G2
Synthetic peptides corresponding to residues 796-812 of human DaP-5 protein.
Protein G
RUO
Antibody
Monoclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit