Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-DARS, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP133689

Catalog No. NBP133689


Add to cart

Description

Description

DARS Polyclonal antibody specifically detects DARS in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin.
Specifications

Specifications

DARS
0.35mg/mL
Western Blot 1:500-1:3000, Immunohistochemistry 10 - 1:500, Immunocytochemistry/Immunofluorescence 1:100-1:1000, Immunoprecipitation 1:10 - 1:500, Immunohistochemistry-Paraffin 1:100-1:1000
Affinity Purified
DARS
Recombinant protein encompassing a sequence within the center region of human DARS. The exact sequence is proprietary.
Immunogen affinity purified
RUO
Primary
1615
Human
Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Unconjugated
0.1M Tris (pH 7.0), 0.1M Glycine and 20% Glycerol with 0.01% Thimerosal
aspartate tRNA ligase 1, cytoplasmic, Aspartate--tRNA ligase, aspartyl-tRNA synthetase, aspartyl-tRNA synthetase, cytoplasmic, aspRS, Cell proliferation-inducing gene 40 protein, cell proliferation-inducing protein 40, DKFZp781B11202, EC 6.1.1.12, MGC111579
Rabbit
IgG
0.1 ml
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Polyclonal
Tested in human. Predicted cross-reactivity based on sequence identity: Xenopus laevis (90%) and Xenopus tropicalis (90%). Not yet tested in any other species.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only