Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-DAX1/NR0B1, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP152822

Catalog No. NBP152822


Add to cart

Description

Description

DAX1/NR0B1 Polyclonal antibody specifically detects DAX1/NR0B1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications

Specifications

DAX1/NR0B1
Unconjugated
PBS and 2% Sucrose with 0.09% Sodium Azide
Purified
NR0B1
Synthetic peptides corresponding to NR0B1(nuclear receptor subfamily 0, group B, member 1) The peptide sequence was selected from the N terminal of NR0B1. Peptide sequence TSSKQTHVAPAAPEARPGGAWWDRSYFAQRPGGKEALPGGRATALLYRCC.
Protein A purified
RUO
Store at -20C. Avoid freeze-thaw cycles.
Polyclonal
Expected identity based on immunogen sequence: Human: 100%.
Human
Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Western Blot 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500
P51843
AHCDSS, AHCH, AHX, DAX1DAX-1, dosage-sensitive sex reversal, DSS-AHC critical region on the X chromosome protein 1, GTD, HHG, NROB1, nuclear hormone receptor, Nuclear receptor DAX-1, nuclear receptor subfamily 0 group B member 1, nuclear receptor subfamily 0, group B, member 1
Rabbit
IgG
100ul
Cancer, GPCR
Primary
190
Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 100μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only