Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NR0B1 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA512270

Catalog No. PIPA512270

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

NROB1contains a DNA-binding domain and acts as a domit-negative regulator of transcription which is mediated by the retinoic acid receptor. This protein also functions as an anti-testis gene by acting antagonistically to Sry. Mutations in the NROB1 gene result in both X-linked congenital adrenal hypoplasia and hypogonadotropic hypogonadism.
Specifications

Specifications

NR0B1
Polyclonal
Unconjugated
NR0B1
Liquid
NR0B1
KLH conjugated synthetic peptide between 304-333 amino acids from the C-terminal region of human DAX1 (NR0B1)
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Western Blot
0.34 mg/mL
PBS with 0.09% sodium azide
P51843
Nuclear receptor subfamily group B member 1; Nuclear receptor DAX-1; DSS-AHC critical region on the X chromosome protein 1; NR0B1; AHC; DAX1
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
190
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit