Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DAZL Rabbit anti-Bovine, Canine, Human, Mouse, Non-human primate, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA523152

Catalog No. PIPA523152


Add to cart

Description

Description

PA5-23152 targets DAZL in WB applications and shows reactivity with Bovine, Canine, Human, mouse, new world monkey and chimpanzee samples.

Protein expression pattern show that DAZL is involved in mitosis and meiosis of human germ cell. The DAZL gene family encodes potential RNA binding proteins that are expressed in prenatal and postnatal germ cells of males and females .The protein is expressed in germ cells of diverse mammalian species. The protein encoded by this gene is localized to the nucleus and cytoplasm of fetal germ cells and to the cytoplasm of developing oocytes. In humans, DAZL is expressed in primordial germ cells, adult testis, and the ovary. It plays a central role during spermatogenesis. Defects in DAZL may cause infertility due to severe oligozoospermia (az) and non obstructive azoospermia. The concentrations of DAZL transcripts were lower in men with spermatogenic failure. Drosophila and mouse mutants that have lost DAZ homologous gene function found to be azoospermic, just as with human azoospermia DAZ. Transposition and amplification of this autosomal gene during primate evolution gave rise to the DAZ gene cluster on the Y chromosome.
Specifications

Specifications

DAZL
Polyclonal
whole serum with 50% glycerol and 0.025% sodium azide
Q92904, Q64368
DAZH, DAZL1, DAZLA, SPGYLA, DAZ homolog, DAZ-like autosomal, SPGY-like-autosomal, deleted in azoospermia-like 1, germline specific RNA binding protein, spermatogenesis gene on the Y-like autosomal
Rabbit
IgG
RUO
Antibody
Polyclonal
Bovine, Canine, Human, Mouse, Non-human primate
Western Blot
Unconjugated
DAZL
Liquid
DAZL
Synthetic peptide (50 RMDETEIRSFFARYG 64) of DAZL protein.
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
13164, 1618, 477055, 530116
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit