Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DC-SIGN+DC-SIGNR Mouse anti-Human, Clone: 120612, R&D Systems™

Mouse Monoclonal Antibody

$119.60 - $839.00

Specifications

Antigen DC-SIGN+DC-SIGNR
Immunogen NIH-3T3 mouse embryonic fibroblast cell line transfected with human DC-SIGNR Accession # Q9H2X3
Clone 120612
Applications Flow Cytometry, Immunohistochemistry, CyTOF
Conjugate Unconjugated
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MAB1621SP
View Documents
R&D Systems
MAB1621SP
25 ug
Each for $119.60
Add to cart
 
MAB1621100
View Documents
R&D Systems
MAB1621100
100 ug
Each for $377.00
Add to cart
 
MAB1621500
View Documents
R&D Systems
MAB1621500
500 ug
Each for $839.00
Add to cart
 
Description

Description

DC-SIGN+DC-SIGNR Monoclonal antibody MAB1621 Specifically detects DC-SIGN+DC-SIGNR in Human samples. It is validated for Flow Cytometry, Immunohistochemistry
Specifications

Specifications

DC-SIGN+DC-SIGNR
120612
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Mouse
IgG2a
30835
RUO
NIH-3T3 mouse embryonic fibroblast cell line transfected with human DC-SIGNR Accession # Q9H2X3
Flow Cytometry, Immunohistochemistry, CyTOF
Purified
Q9H2X3
Monoclonal
Primary
Recognizes both human DC-SIGN and human DC-SIGNR on transfected cells. Does not react with parental mouse cells or irrelevant transfectants.
Protein A or G purified from hybridoma culture supernatant
SDS
Product Certifications

For Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit