Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CLEC4B Rat anti-Mouse, Clone: 349214, Invitrogen™

Rat Monoclonal Antibody

Manufacturer:  Invitrogen MA524089

Catalog No. PIMA524089


Add to cart

Description

Description

Reconstitute at 0.5 mg/mL in sterile PBS.

Specifications

Specifications

CLEC4B
Monoclonal
0.5 mg/mL
PBS with 5% trehalose and No Preservative
Lyophilized
Clec4b1
Mouse myeloma cell line NS0-derived recombit mouse DCAR/CLEC4B Gln42-Leu209
Protein A/G
RUO
Antibody
Monoclonal
Mouse
Flow Cytometry
349214
Unconjugated
Clec4b1
Aplra2, CLEC4B
Rat
IgG2A
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
69810
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit