Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DSCC1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA524625
Description
DSCC1 Polyclonal antibody specifically detects DSCC1 in Human samples. It is validated for Western Blot.Specifications
DSCC1 | |
Polyclonal | |
PBS with 0.09% sodium azide | |
Q9BVC3 | |
DSCC1 | |
KLH conjugated synthetic peptide between 311-339 amino acids from the C-terminal region of human DCC1 | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
DSCC1 | |
2010006I05Rik; 2600005O03Rik; DCC1; defective in sister chromatid cohesion 1 homolog; defective in sister chromatid cohesion 1 homolog (S. cerevisiae); defective in sister chromatid cohesion protein 1 homolog; DNA replication and sister chromatid cohesion 1; DSCC1; RGD1561749; Sister chromatid cohesion protein DCC1; UNQ9337/PRO34008 | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
79075 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction