Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Artemis Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA526741

Catalog No. PIPA526741

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

DCLRE1C is a nuclear protein that is involved in V(D)J recombination and DNA repair. The protein has single-strand-specific 5 ft.-3 ft. exonuclease activity; it also exhibits endonuclease activity on 5 ft. and 3 ft. overhangs and hairpins when complexed with protein kinase, DNA-activated, catalytic polypeptide.
Specifications

Specifications

Artemis
Polyclonal
PBS with 0.09% sodium azide
Q96SD1, Q8K4J0
A-SCID, DCLREC1C, RS-SCID, SCIDA, SNM1C, hSNM1C, DNA cross-link repair 1C (PSO2 homolog, S. cerevisiae), DNA cross-link repair 1C protein, PSO2 homolog, SNM1 homolog C, SNM1-like protein, protein artemis, severe combined immunodeficiency, type a (Athabascan)
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
227525, 64421
Immunohistochemistry, Western Blot
Unconjugated
DCLRE1C
Liquid
DCLRE1C
KLH conjugated synthetic peptide between 21-50 amino acids from the N-terminal region of human DCLRE1C
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit