Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DCP1B Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA526885

Catalog No. PIPA526885

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

DCP1B is a core component of the mRNA decapping complex, a key factor in the regulation of mRNA decay.
Specifications

Specifications

DCP1B
Polyclonal
PBS with 0.09% sodium azide
Q8IZD4
DCP1, hDcp1b, decapping enzyme hDcp1b, mRNA-decapping enzyme 1B
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
196513
Flow Cytometry, Western Blot
Unconjugated
DCP1B
Liquid
DCP1B
KLH conjugated synthetic peptide between 261-290 amino acids from the central region of human DCP1B
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit