Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

dCMP deaminase, Mouse, Polyclonal Antibody, Abnova™

Mouse polyclonal antibody raised against a full-length human DCTD protein.

Manufacturer:  Abnova Corporation H00001635B01

Catalog No. 89-005-230


Add to cart

Description

Description

The protein encoded by this gene catalyzes the deamination of dCMP to dUMP, the nucleotide substrate for thymidylate synthase. The encoded protein is allosterically activated by dCTP and inhibited by dTTP, and is found as a homohexamer. This protein uses zinc as a cofactor for its activity. Two transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq

Sequence: MVGGGQPCGPNMSEVSCKKRDDYLEWPEYFMAVAFLSAQRSKDPNSQVGACIVNSENKIVGIGYNGMPNGCSDDVLPWRRTAENKLDTKYPYVCHAELNAIMNKNSTDVKGCSMYVALFPCNECAKLIIQAGIKEVIFMSDKYHDSDEATAARLLFNMAGVTFRKFIPKCSKIVIDFDSINSRPSQKLQ
Specifications

Specifications

dCMP deaminase
Unconjugated
No additive
NM_001012732.1
MGC111062
Mouse
50 uL
Endocrine/Metabolism
Y
Polyclonal
Human
Western Blot
Mouse polyclonal antibody raised against a full-length human DCTD protein.
DCTD
Serum
DCTD
DCTD (NP_001012750.1, 1 a.a. ~ 189 a.a) full-length human protein.
RUO
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Primary
1635
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit