Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DcTRAILR2 Rat anti-Mouse, Clone: 263310, Invitrogen™

Rat Monoclonal Antibody

Manufacturer:  Invitrogen MA523964

Catalog No. PIMA523964


Add to cart

Description

Description

In direct ELISAs and Western blots, no cross-reactivity with recombit mouse (rm) 4-1BB, rmBAFF R, rmCD27, rmCD30, rmCD40, rmDcTRAIL R1, rmDR3, rmEDAR, rmFas, rmGITR, rmHVEM, rmLT beta R, rmNGF R, rmOPG, rmOX40, rmRANK, rmTNF RI, and rmTNF RII is observed. Reconstitute at 0.5 mg/mL in sterile PBS.

Specifications

Specifications

DcTRAILR2
Monoclonal
0.5 mg/mL
PBS with 5% trehalose and No Preservative
Q9ER62
SOBa, TNFRH2, TNFRSF22
Rat
IgG2B
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
79202
Western Blot
263310
Unconjugated
Tnfrsf22
Lyophilized
TNFRSF22
Mouse myeloma cell line NS0-derived recombit mouse DcTRAIL R2/TNFRSF22 Ala39-Arg175
Protein A/G
RUO
Antibody
Monoclonal
Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit