Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DCXR Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA514345

Catalog No. PIPA514345

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

DCXR is an enzyme that has both diacetyl reductase and L-xylulose reductase activities.
Specifications

Specifications

DCXR
Polyclonal
PBS with 0.09% sodium azide
Q7Z4W1, Q91X52
Sperm surface protein P34H; Carbonyl reductase II; Kidney dicarbonyl reductase; kiDCR; Dicarbonyl/L-xylulose reductase; L-xylulose reductase; XR; DCXR
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
51181, 67880
Flow Cytometry, Immunohistochemistry, Western Blot
Unconjugated
DCXR
Liquid
DCXR
KLH conjugated synthetic peptide between 79-106 amino acids from the central region of human DCXR
Ammonium sulfate precipitation, Size-exclusion - Dialysis
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit