Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DDB1 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA521282

Catalog No. PIPA521282


Add to cart

Description

Description

PA5-21282 targets DDB1 in ICC, IF, IHC (P), and WB applications and shows reactivity with Human and mouse samples. The PA5-21282 immunogen is a recombit fragment corresponding to a region within amino acids 749 and 1140 of DDB1 (Uniprot IDNo.Q16531). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.

Specifications

Specifications

DDB1
Polyclonal
Unconjugated
DDB1
Liquid
DDB1
Recombit fragment corresponding to a region within amino acids 749 and 1140 of DDB1 (Uniprot IDNo.Q16531)
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
0.81 mg/mL
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
Q16531, Q3U1J4
DDBA, UV-DDB1, XAP1, XPCE, XPE, XPE-BF, DDB p127 subunit, DNA damage-binding protein 1, DNA damage-binding protein a, HBV X-associated protein 1, UV-DDB 1, UV-damaged DNA-binding factor, UV-damaged DNA-binding protein 1, XAP-1, XPE-binding factor, damage-specific DNA-binding protein 1, xeroderma pigmentosum group E-complementing protein
Rabbit
IgG
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
13194, 1642
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit