Learn More
DDOST, Mouse, Clone: 2D7, Abnova™
Mouse monoclonal antibody raised against a partial recombinant DDOST.
Supplier: Abnova Corporation H00001650M06
Description
This gene encodes a component of the oligosaccharyltransferase complex which catalyzes the transfer of high-mannose oligosaccharides to asparagine residues on nascent polypeptides in the lumen of the rough endoplasmic reticulum. The protein complex co-purifies with ribosomes. The product of this gene is also implicated in the processing of advanced glycation endproducts (AGEs), which form from nonenzymatic reactions between sugars and proteins or lipids and are associated with aging and hyperglycemia. [provided by RefSeq
Sequence: TDLVEYSIVIQQLSNGKWVPFDGDDIQLEFVRIDPFVRTFLKKKGGKYSVQFKLPDVYGVFQFKVDYNRLGYTHLYSSTQVSVRPLQHTQYERFIPSAYPSpecifications
DDOST | |
Monoclonal | |
Unconjugated | |
PBS with no preservative; pH 7.4 | |
NM_005216 | |
DDOST | |
DDOST (NP_005207, 328 a.a. ∼ 427 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2a κ |
ELISA, Immunofluorescence, Western Blot | |
2D7 | |
Mouse monoclonal antibody raised against a partial recombinant DDOST. | |
DDOST | |
AGE-R1/KIAA0115/MGC2191/OK/SW-cl.45/OST/OST48/WBP1 | |
Mouse | |
Affinity chromatography | |
RUO | |
1650 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.