Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DDX1 Mouse anti-Human, Unlabeled, Clone: 22, BD

Mouse Monoclonal Antibody

Manufacturer:  BD Biosciences 611206

 View more versions of this product

Catalog No. BDB611206


Add to cart

Description

Description

DEAD box proteins, a family of putative RNA helicases, are characterized by eight conserved amino acid motifs that are arranged in a core region as found in the prototypical member of the family, eIF-4A. The family's name is derived from the amino acid sequence Asp-Glu-Ala-Asp (DEAD) that is located within the ATP hydrolysis motif. DEAD box proteins have been implicated in translation initiation and RNA splicing, degradation, and stability. The DEAD box protein, DDX1, was identified in retinoblastoma cell lines, where it is coamplified with MYCN, a member of the MYC transcription factor family. DDX1 is expressed at elevated levels in fetal tissues of neural origin (retina and brain). It is both cytoplasmic and nuclear in DDX1-amplified neuroblastoma and retinoblastoma cell lines. However, it localizes specifically to the nucleus in non-amplified cell lines. Thus, DDX1 is thought to be involved in nuclear RNA metabolism. In addition, the decrease in mean disease-free survival of neuroblastoma patients exhibiting coamplification of DDX1 and MYCN as compared to those exhibiting MYCN amplification indicates a possible role for DDX1 in cancer formation in neural tissues.

Host Species: Mouse
Clone: 22
Isotype: IgG1
Species Reactivity: Human
Immunogen: Human DDX1 aa. 351-544
Formula Weight [Chemical]: 82kDa

Immunofluorescence, Western Blotting

Specifications

Specifications

DDX1
22
Unconjugated
Affinity Purified
Human DDX1 aa. 351-544
50μg
Cell Biology
Monoclonal
Immunofluorescence, Western Blot
250μg/mL
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Murine
IgG1
RUO
Primary
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only.