Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DDX11 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA523956

Catalog No. PIPA523956


Add to cart

Description

Description

DEAD box proteins, characterized by the conserved motifAsp-Glu-Ala-Asp (DEAD), are putative RNA helicases. They areimplicated in a number of cellular processes involving alterationof RNA secondary structure such as translation initiation, nuclearand mitochondrial splicing, and ribosome and spliceosome assembly.Based on their distribution patterns, some members of this familyare believed to be involved in embryogenesis, spermatogenesis, andcellular growth and division. DDX11 encodes a DEAD box protein,which is an enzyme that possesses both ATPase and DNA helicaseactivities. DDX11 is a homolog of the yeast CHL1 gene, and mayfunction to maintain chromosome transmission fidelity and genomestability.
Specifications

Specifications

DDX11
Polyclonal
Unconjugated
DDX11
Liquid
DDX11
KLH conjugated synthetic peptide between 656-683 amino acids from the central region of human DDX11
Protein A, Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Flow Cytometry, Western Blot
0.5 mg/mL
PBS with 0.09% sodium azide
Q96FC9, Q6AXC6
CHL1, CHLR1, KRG2, CHL1-like helicase homolog, CHL1-related helicase gene-1, CHL1-related protein 1, DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 11 (CHL1-like helicase homolog, S. cerevisiae), DEAD/H box protein 11, KRG-2, hCHLR1, keratinocyte growth factor-regulated gene 2 prote
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
1663, 320209
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit