Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DDX23 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA524792

Catalog No. PIPA524792


Add to cart

Description

Description

DDX23 encodes a member of the DEAD box protein family. DEAD box proteins, characterized by the conserved motif Asp-Glu-Ala-Asp (DEAD), are putative RNA helicases. They are implicated in a number of cellular processes involving alteration of RNA secondary structure, such as translation initiation, nuclear and mitochondrial splicing, and ribosome and spliceosome assembly. Based on their distribution patterns, some members of this family are believed to be involved in embryogenesis, spermatogenesis, and cellular growth and division. The protein is a component of the U5 snRNP complex; it may facilitate conformational changes in the spliceosome during nuclear pre-mRNA splicing.
Specifications

Specifications

DDX23
Polyclonal
PBS with 0.09% sodium azide
Q9BUQ8
PRPF28, U5-100K, U5-100KD, prp28, 100 kDa U5 snRNP-specific protein, DEAD box protein 23, PRP28 homolog, yeast, PRP28p homolog, U5 snRNP 100 kD protein, probable ATP-dependent RNA helicase DDX23
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
9416
Flow Cytometry, Immunohistochemistry, Western Blot
Unconjugated
DDX23
Liquid
DDX23
KLH conjugated synthetic peptide between 248-277 amino acids from the central region of human DDX23
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit