Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DDX4 Rabbit anti-Canine, Equine, Human, Mouse, Non-human primate, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA523378

Catalog No. PIPA523378


Add to cart

Description

Description

DDX4 belongs to the DEAD box helicase family or DDX4/VASA subfamily. DEAD box proteins are characterized by the conserved Asp-Glu-Ala-Asp (DEAD) motif and are putative RNA helicases. This protein has a glycine-rich N terminus with multiple repeats of an RGG motif, which is believed to function in RNA binding. They have a role in a number of cellular processes involving alteration of RNA secondary structure such as translation initiation, nuclear and mitochondrial splicing, and ribosome and spliceosome assembly. The members of this family are involved in embryogenesis, spermatogenesis, and cellular growth and division. This gene encodes a DEAD box protein, which is a homolog of VASA proteins in Drosophila. It is specifically expressed in the germ cell lineage in both sexes and functions in germ cell development. It is required during gametogenesis in the adult and in the specification of the germ cell lineage during embryogenesis. This is conserved in invertebrate and vertebrate species.
Specifications

Specifications

DDX4
Polyclonal
Unconjugated
DDX4
Liquid
DDX4
Synthetic peptide (711 SSQAPNPVDDESWD 724) of DDX4 protein.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Canine, Equine, Human, Mouse, Non-human primate
Immunohistochemistry (Paraffin)
0.5 mg/mL
PBS with 0.05% BSA and 0.05% sodium azide
Q9NQI0, Q61496
VASA, DEAD box protein 4, DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 4, probable ATP-dependent RNA helicase DDX4, vasa homolog
Rabbit
IgG
100 μg
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
100051760, 13206, 487211, 54514
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit