Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DDX5 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA514366

Catalog No. PIPA514366

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

This antibody is predicted to react with mouse and non-human primate based on sequence homology.

DDX5 is putative RNA helicases. The protein is implicated in a number of cellular processes involving alteration of RNA secondary structure, such as translation initiation, nuclear and mitochondrial splicing, and ribosome and spliceosome assembly. Based on their distribution patterns, some members of this family are believed to be involved in embryogenesis, spermatogenesis, and cellular growth and division. This protein is a RNA-dependent ATPase, and also a proliferation-associated nuclear antigen, specifically reacting with the simian virus 40 tumor antigen.
Specifications

Specifications

DDX5
Polyclonal
PBS with 0.09% sodium azide
P17844
Probable ATP-dependent RNA helicase DDX5; DEAD box protein 5; RNA helicase p68 ; HLR1; HELR; G17P1; DDX5
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
1655
Immunohistochemistry, Western Blot
Unconjugated
DDX5
Liquid
DDX5
KLH conjugated synthetic peptide between 491-518 amino acids from the C-terminal region of human DDX5
Ammonium sulfate precipitation, Size-exclusion - Dialysis
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit