Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DEC1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA523502

Catalog No. PIPA523502


Add to cart

Description

Description

Dec1, which is also known as Stra13 or Sharp2, is a basic helix-loop-helix (bHLH) transcription factor that has been found to play an important role in circadian system. Dec1 contains one basic helix-loop-helix (bHLH) domain, one Orange domain and is reported to be expressed in Treg cells. It is also reported that Dec1 is required for long-term maintece of Treg cells after adoptive transfer to suppress effector T cell-mediated inflammation. Dec1 positively regulates CD25 expression in association with Runx 1.
Specifications

Specifications

DEC1
Polyclonal
Unconjugated
DEC1
Liquid
DEC1
Synthetic peptide (34 YQVYKSRRGIKRSEDSKE 51) of Dec1 protein.
Protein A
RUO
Antibody
Polyclonal
Human, Mouse, Rat
Immunohistochemistry (Paraffin), Western Blot
1.0 mg/mL
PBS with 0.05% sodium azide
Q9P2X7
CTS9, candidate tumor suppressor CTS9
Rabbit
IgG
100 μg
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
50514
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit