Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-DEC2/SHARP1, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

$119.60 - $314.00

Specifications

Host Species Rabbit
Isotype IgG
Immunogen A synthetic peptide made to an internal portion of the human DEC2/SHARP1 inbetween residues 1-75 [UniProt Q9C0J9].
Antigen DEC2/SHARP1
Monoclonal or Polyclonal Polyclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP119613SS
View Documents
Novus Biologicals
NBP119613SS
0.025 mg
Each for $119.60
Add to cart
 
NBP119613
View Documents
Novus Biologicals
NBP119613
0.1 mg
Each for $314.00
Add to cart
 
Description

Description

DEC2/SHARP1 Polyclonal antibody specifically detects DEC2/SHARP1 in Human, Mouse, Rat samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications

Specifications

Rabbit
A synthetic peptide made to an internal portion of the human DEC2/SHARP1 inbetween residues 1-75 [UniProt Q9C0J9].
Polyclonal
Primary
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Q9C0J9
PBS with 0.02% Sodium Azide
IgG
DEC2/SHARP1
Affinity Purified
RUO
Human, Mouse, Rat. Immunogen has 100% homology to bovine and canine.
79365
1.0 mg/mL
SDS
Product Certifications

For Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit