Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Decorin Mouse anti-Human, Clone: 115402, R&D Systems™

Mouse Monoclonal Antibody

$125.00 - $839.00

Specifications

Antigen Decorin
Immunogen S. frugiperda insect ovarian cell line Sf 21-derived recombinant human Decorin Gly17-Lys172 Accession # NP_598013.1
Clone 115402
Applications Western Blot, Immunohistochemistry
Conjugate Unconjugated
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MAB143SP
View Documents
R&D Systems
MAB143SP
25 ug
Each for $125.00
Add to cart
 
MAB143100
View Documents
R&D Systems
MAB143100
100 ug
Each for $377.00
Add to cart
 
MAB143500
View Documents
R&D Systems
MAB143500
500 ug
Each for $839.00
Add to cart
 
Description

Description

Decorin Monoclonal antibody MAB143 Specifically detects Decorin in Human samples. It is validated for Immunohistochemistry, Simple Western, Western Blotting
Specifications

Specifications

Decorin
115402
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Mouse
IgG1
1634
RUO
S. frugiperda insect ovarian cell line Sf 21-derived recombinant human Decorin Gly17-Lys172 Accession # NP_598013.1
Western Blot, Immunohistochemistry
Purified
NP_598013.1
Monoclonal
Primary
Detects human Decorin in direct ELISAs and Western blots. In direct ELISAs, this antibody does not cross-react with recombinant mouse Decorin.
Protein A or G purified from hybridoma culture supernatant
SDS
Product Certifications

For Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit