Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DECR2, Rabbit, Monoclonal Antibody, Abnova™
Rabbit monoclonal antibody raised against a human DECR2 peptide using ARM Technology.
Supplier: Abnova Corporation H00026063K
Includes: Up to three rabbit IgG clones of 1mg each will be delivered to customer.
Description
Library Construction: Nonfusion antibody library from rabbit spleen (ARM Technology)Expression: Overexpression vector and transfection into 293H cell line
Order Info
- Customer may provide cell or tissue lysate for antibody screening.
- Rabbit monoclonal antibody generated by ARM technology is amenable to antibody engineering including F(ab)2, IgG, scFv and different Fc and non-Fc conjugates per customer request.
Specifications
DECR2 | |
Monoclonal | |
Unconjugated | |
In 1X PBS, pH 7.4 | |
DECR2 | |
DECR2 | |
A synthetic peptide of human DECR2 | |
RUO | |
26063 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
IgG |
ELISA, Western Blot | |
1mg | |
2,4-dienoyl CoA reductase 2, peroxisomal | |
DECR2 | |
PDCR, SDR17C1 | |
Rabbit | |
1 mg x up to 3 Abs | |
Primary | |
Human | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction