Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DEDAF Rabbit anti-Human, Mouse, Rat, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Supplier: MilliporeSigma AB3637
Description
This Anti-DEDAF Antibody is validated for use in western blotting for the detection of DEDAF.
Specifications
DEDAF | |
Polyclonal | |
Purified IgG by immunoaffinity chromatography. Liquid in PBS containing 0.02% sodium azide. | |
Rabbit | |
100 μg | |
NM_012234.4 | |
2-8°C in undiluted aliquots for up to 12 months |
Western Blot | |
Unconjugated | |
RYBP; DEDAF; YEAF1; AAP1; APAP-1 | |
Rabbit anti-DEDAF polyclonal antibody was raised against a synthetic peptide corresponding to amino acids 215 to 228 of human-DEDAF. This sequence is identical to the mouse sequence. | |
Primary | |
Human, Mouse, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction