Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

OPRD1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA127970

Catalog No. PIPA127970


Add to cart

Description

Description

PA1-27970 detects delta Opioid receptor from rat and mouse samples. Expected to cross react with human samples due to 100% sequence homology. PA1-27970 has been successfully used in immunocytochemistry applications. The PA1-27970 immunogen is synthetic peptide: TACTPSDGPGGGAAA, corresponding to amino acids 358-372 of Rat Delta Opioid Receptor. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.

Specifications

Specifications

OPRD1
Polyclonal
whole serum with 0.05% sodium azide
P41143, P32300, P33533
OPRD1
synthetic peptide TACTPSDGPGGGAAA, corresponding to amino acids 358-372 of Rat Delta Opioid Receptor.
50 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
18386, 24613, 4985
Immunocytochemistry, Immunofluorescence
Unconjugated
OPRD1
Liquid
Rabbit
IgG
RUO
Antibody
Polyclonal
Human, Mouse, Rat
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit