Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dengue Virus Type 1-4 Envelope, clone: DE-2, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA171251

Catalog No. MA171251

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Dengue fever and dengue hemorrhagic fever are acute febrile diseases, found in the tropics and Africa. Caused by one of four closely related virus serotypes of the genus Flavivirus, family Flaviviridae, each serotype is sufficiently different that there is no cross-protection and epidemics caused by multiple serotypes (hyperendemicity) can occur. Dengue is transmitted to humans by the Aedes aegypti (rarely Aedes albopictus) mosquito, which feeds during the day.

MA1-71251 may also be used in a indirect immunofluorescence assay for the detection of dengue virus antigens in infected cells. It has been tested in other ELISA formats including the dengue virus antigen capture ELISA, dengue virus IgM capture ELISA and the dengue virus IgG capture ELISA. This monoclonal antibody is not applicable for western blot immunoassays.
Specifications

Specifications

Dengue Virus Type 1-4 Envelope
Monoclonal
Unconjugated
Supernatant
Murine
IgG2a, κ
RUO
Antibody
Monoclonal
ELISA, Immunofluorescence
DE-2
tissue culture supernatant with 10mM sodium azide
Dengue envelope, Dengue Virus 2 Envelope, Dengue 2 envelope, Dengue pan
Envelope glycoprotein of dengue virus serotype 1, 2, 3 and 4.
1mL
Store at 4°C
Primary
Virus
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit