Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TSPYL2 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA14261

Catalog No. PA14261


Add to cart

Description

Description

PA1-4261 detects the DENTT protein in human samples. PA1-4261 has been successfully used Western blot procedures. PA1-4261 immunizing peptide corresponds to amino acid residues 489-503 of the human DENTT protein.

A novel gene, differentially expressed nucleolar TGF-beta1 target (DENTT), is expressed in several normal human tissues, with the highest level of expression in brain. It is involved in TGF-beta1-mediated responses and has distinct domains for direction to different subnuclear locations.
Specifications

Specifications

TSPYL2
Polyclonal
Unconjugated
TSPYL2
Liquid
TSPYL2
Synthetic peptide: N(489) N E T T D N N E S A D D H E(503)
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Western Blot
1 mg/mL
PBS with 0.02% sodium azide
Q9H2G4
RP1-290F12.2, CDA1, CINAP, CTCL, DENTT, HRIHFB2216, NP79, SE204, TSPX
Rabbit
IgG
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
64061
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit