Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DEPDC1 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA525343

Catalog No. PIPA525343


Add to cart

Specifications

Specifications

DEPDC1
Polyclonal
PBS with 0.09% sodium azide
Q5TB30
DEP.8, DEPDC1-V2, DEPDC1A, SDP35, 5830484J08Rik, DEP domain-containing protein 1A, cell cycle control protein SDP35
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
55635
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
DEPDC1
Liquid
DEPDC1
KLH conjugated synthetic peptide between 10-38 amino acids from the N-terminal region of human DEPDC1
Protein A, Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit