Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DEPTOR/DEPDC6 Antibody, DyLight 650, Novus Biologicals™
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP149674C
Description
DEPTOR/DEPDC6 Polyclonal specifically detects DEPTOR/DEPDC6 in Human, Mouse, Rat, Bovine samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
DEPTOR/DEPDC6 | |
Polyclonal | |
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin | |
Q8TB45 | |
DEPTOR | |
A genomic peptide made to an internal region of the human DEPTOR protein (within residues 10-200). [Swiss-Prot: Q8TB45] | |
0.1 mL | |
Cancer, Signal Transduction | |
64798 | |
Human, Mouse, Rat, Bovine | |
IgG |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
DyLight 650 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
DEP domain containing 6, DEP domain containing mTOR interacting protein, DEP domain containing MTOR-interacting protein, DEP domain-containing mTOR-interacting protein, DEP domain-containing protein 6, DEP.6, DEPDC6FLJ12341, DKFZp564B1778, FLJ12428, FLJ13854 | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
Human and mouse. Predicted to react with bovine based on 100% sequence homology. | |
Store at 4C in the dark. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction