Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-DEPTOR/DEPDC6, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

$119.60 - $293.00

Specifications

Host Species Rabbit
Isotype IgG
Immunogen A portion of amino acids 230-300 of human DEPOR/DEPDC6 was used as the immunogen.
Antigen DEPTOR/DEPDC6
Monoclonal or Polyclonal Polyclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP227083SS
View Documents
Novus Biologicals
NBP227083SS
0.025 mg
Each for $119.60
Add to cart
 
NBP227083
View Documents
Novus Biologicals
NBP227083
0.1 mg
Each for $293.00
Add to cart
 
Description

Description

DEPTOR/DEPDC6 Polyclonal antibody specifically detects DEPTOR/DEPDC6 in Human, Rat, Equine, Primate samples. It is validated for Western Blot.
Specifications

Specifications

Rabbit
A portion of amino acids 230-300 of human DEPOR/DEPDC6 was used as the immunogen.
Polyclonal
Primary
Western Blot
Q8TB45
PBS, 0.05% BSA with 0.05% Sodium Azide
IgG
DEPTOR/DEPDC6
Purified
RUO
The amino acid sequence used as immunogen is 100% homologous in human, monkey, chimpanzee, horse and rat 93% homologous in mouse andcow, and 87% homologous in chicken.
64798
0.5 mg/mL
SDS
Product Certifications

For Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit