Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Desmin Rabbit anti-Human, Mouse, Rat, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA137556

Catalog No. 01-673-363


Add to cart

Description

Description

This antibody is predicted to react with bovine, chicken, canine, amphibian, porcine, rabbit and rat based on sequence homology. A suggested positive control is skeletal muscle or sarcoma.

Desmin is a 53 kDa intermediate filament protein expressed in cardiac, skeletal, and smooth muscle cells, as well as lung tissue. In skeletal and cardiac muscle cells, desmin is localized at the Z-disk region and the intercalated disk. In smooth muscle tissue, desmin is expressed throughout the cells. During development, levels of desmin expression vary, being present at low levels in the early stages of myoblast replication and increasing during terminal differentiation. Unlike vimentin, desmin is expressed in both differentiating and mature muscle cells. Inhibiting desmin expression blocks myoblast fusion and myotube formation, suggesting that desmin is crucial during myogenesis.
Specifications

Specifications

Desmin
Polyclonal
PBS with 1% BSA and 0.1% sodium azide
P17661, P48675, P31001
CMD1I, CSM2, DES, FLJ12025
Rabbit
IgG
1 mL
-20° C, Avoid Freeze/Thaw Cycles
Primary
13346, 64362, 1674
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin)
Unconjugated
DES
Liquid
DES
Synthetic peptide mapping near the C-terminus of human desmin
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit