Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Destrin Rabbit anti-Human, Mouse, Rat, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA124936

Catalog No. PIPA124936


Add to cart

Description

Description

PA1-24936 detects Destrin from human, mouse, and rat samples. PA1-24936 has been successfully used in Western blot procedures. The PA1-24936 immunogen is a synthetic peptide, conjugated to KLH, corresponding to residues 137-150 of Destrin, with an N-terminal added lysine.

Specifications

Specifications

Destrin
Polyclonal
Unconjugated
DSTN
Liquid
DSTN
Synthetic peptide, conjugated to KLH, corresponding to residues 137-150 of Destrin, with an N-terminal added lysine.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat
Western Blot
0.9 mg/mL
0.01M PBS with 1% BSA and 15mM sodium azide; pH 7.4
P60981, Q9R0P5, Q7M0E3
RP23-77A2.1, 2610043P17Rik, ADF, AU042046, Dsn, corn1, sID
Rabbit
IgG
50 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
11034, 502674, 56431
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit