Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DFF45 Mouse anti-Human, Unlabeled, Clone: 19, BD

Mouse Monoclonal Antibody

Manufacturer:  BD Biosciences 611036

Catalog No. BDB611036


Add to cart

Description

Description

Apoptosis, a selective process of genetically programmed cell death, occurs during normal cellular differentiation and development of multicellular organisms. Apoptotic cells are characterized by loss of cell volume, plasma membrane blebbing, nuclear condensation, chromatin aggregation, and endonucleocytic degradation of DNA into nucleosomal fragments. Caspase-3 (CPP32, Yama, apopain) is the most extensively studied apoptotic protein. An apoptotic signal induces the intracellular cleavage of caspase-3 from an inactive proform to an active fragment. The active form of caspase-3 cleaves several other apoptotic proteins including DNA Fragmentation Factor (DFF). DFF is thought to be either directly or indirectly responsible for the DNA laddering that is a hallmark of apoptosis. It is a heterodimer of 40 kDa (DFF40) and 45 kDa (DFF45) subunits. Cleavage of DFF45 into three fragments is a prerequisite for DFF-mediated DNAfragmentation. Identity of the DFF fragment that mediates DNA laddering remains unclear. However, it is clear that DFF functions downstream of caspase-3 and is essential for the characteristic fragmentation of DNA during apoptosis.
Specifications

Specifications

DFF45
19
Unconjugated
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Murine
IgG1
RUO
Store undiluted at -20°C.
Monoclonal
Western Blot
250μg/mL
DNA Fragmentation Factor-45
Affinity Purified
Human DFF45 aa. 105-305
50μg
Cell Biology
Primary
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit