Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ICAD Rabbit anti-Mouse, Polyclonal, Invitrogen™

Catalog No. PIPA129166
Change view
Click to view available options
Quantity:
500 μL
Catalog No. Quantity
PIPA129166 500 μL
1 options

Catalog No. PIPA129166

Supplier: Thermo Scientific PA129166

Rabbit Polyclonal Antibody

PA1-29167 detects ERK2 from mouse, rat, human samples. PA1-29167 has been successfully used in Western blot applications. The PA1-29167 immunogen is: Synthetic peptide corresponding to the N-terminus of human Erk2 protein.

ICAD is the inhibitor of caspase-3-activated DNase which is a substrate that controls nuclear apoptosis. ICAD has two isoforms: a functional isoform of 45kDa, ICAD-L/DNA fragmentation factor (DFF) 45; and a 35kDa isoform, ICAD-S/DFF35. Although both ICAD-L and ICAD-S can bind and inhibit CAD, only ICAD-L was reported to be functional. ICAD is cleaved to be inactivated and allow caspase-activated DNase (CAD) to execute nuclear internucleosomal apoptotic DNA fragmentation. In non-apoptotic cells, CAD is complexed with its inhibitor, ICAD. The activation of the CAD/ICAD complex occurs through the caspase 3-mediated cleavage of ICAD at residues 117 and 224, which results in three ICAD fragments that are then released from CAD. In addition to its DNase inhibitory activity, ICAD acts as a CAD specific folding chaperone. There are recent reports that ICAD is a potential target for restoring a normal apoptotic signal transduction pathway in colon and brain cancer cells. Two alternatively spliced transcript variants encoding distinct isoforms have been found for the ICAD gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ICAD
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Gene Dffa
Gene Accession No. O54786
Gene Alias RP23-121D17.3, A330085O09Rik, DFF35, Dff45, ICAD, ICAD-L, ICAD-S
Gene Symbols DFFA
Host Species Rabbit
Immunogen Synthetic peptide - near the C terminus of mouse DFF45/ICAD
Purification Method Protein A
Quantity 500 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 13347
Target Species Mouse
Content And Storage 4° C, do not freeze
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.