Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DFFB Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA526840

Catalog No. PIPA526840

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Apoptosis is related to many diseases and induced by a family of cell death receptors and their ligands. Cell death signals are transduced by death domain containing adapter molecules and members of the caspase family of proteases. These death signals finally cause the degradation of chromosomal DNA by activated DNase. A mouse DNase that causes DNA fragmentation was identified recently and designated CAD (for caspase activated deoxyribonuclease). The human homologue of mouse CAD was more recently identified by three groups independently and termed CPAN, DFF40, and human CAD, respectively. DFF45/ICAD is the inhibitory protein of DFF40/CAD and forms complex with DFF40/CAD. Upon cleavage of DFF45/ICAD by activated caspase, DFF40/CAD is released and activated and eventually causes the degradation of DNA in the nuclei. Activation of DFF40/CAD, which causes DNA degradation, is the hallmark of apoptotic cell death.
Specifications

Specifications

DFFB
Polyclonal
PBS with 0.09% sodium azide
O76075
CAD, CPAN, DFF-40, DFF2, DFF40, DNA fragmentation factor 40 kDa subunit, DNA fragmentation factor subunit beta, caspase-activated DNase, caspase-activated deoxyribonuclease, caspase-activated nuclease
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
1677
Flow Cytometry, Immunohistochemistry, Western Blot
Unconjugated
DFFB
Liquid
DFFB
KLH conjugated synthetic peptide between 1-30 amino acids from the N-terminal region of human DFFB
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit