Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ anti-DFHR Clone: 2B10,
Mouse Monoclonal Antibody
Supplier: MilliporeSigma™ ST1711100UG
Description
Specifically detects DFHR Clone: 2B10 in Human, Rat samples, and it is validated for ELISA, Immunocytochemistry, Immuno Blotting
Dihydrofolate reductase converts dihydrofolate into tetrahydrofolate, a methyl group shuttle required for the de novo synthesis of purines, thymidylic acid, and certain amino acids. While the functional dihydrofolate reductase gene has been mapped to chromosome 5, multiple intronless processed pseudogenes or dihydrofolate reductase-like genes have been identified on separate chromosomes. Dihydrofolate reductase deficiency has been linked to megaloblastic anemia.Specifications
DFHR | |
Monoclonal | |
Unconjugated | |
Mouse | |
100 μg | |
Primary | |
Human, Rat | |
Purified |
ELISA, Immunoblot, Immunoblot, Immunocytochemistry | |
2B10 | |
In PBS, pH 7.2. | |
A recombinant polypeptide corresponding to amino acids of 88-188 human DHFR, expressed as a GST fusion protein | |
RUO | |
Recognizes the ∽20-22kDa DHFR protein in HeLa and PC-12 cells. | |
−20°C | |
IgG2a |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction