Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DGAT2 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 3 publications
Supplier: Novus Biologicals NBP171701SS
Description
DGAT2 Polyclonal specifically detects DGAT2 in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
DGAT2 | |
Polyclonal | |
Unconjugated | |
PBS pH7.4 with 0.05% Sodium Azide | |
diacylglycerol O-acyltransferase 2, diacylglycerol O-acyltransferase homolog 2, diacylglycerol O-acyltransferase homolog 2 (mouse), diacylglycerol O-acyltransferase-like protein 2, Diglyceride acyltransferase 2, DKFZp686A15125, EC 2.3.1, EC 2.3.1.20, GS1999FULL | |
Rabbit | |
Affinity Purified | |
RUO | |
84649 | |
Human, Mouse, Rat | |
IgG |
Western Blot | |
1 mg/mL | |
Western Blot 1 ug/ml | |
Q96PD7, Q9DCV3 | |
DGAT2 | |
A synthetic peptide made to an internal region of the human DGAT2 protein (within residues 200-300). [Swiss-Prot Q96PD7] | |
0.025 mL | |
Primary | |
Human and mouse. Immunogen has 100% identity to rat, bovine and porcine, and 88% identity to Xenopus. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction